missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perphenazine 97.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 97°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 403.97 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Perphenazine |
Sécurité et manipulation
| EINECSNumber | (9)-0456 |
| RTECSNumber | TL7175000 |
| TSCA | No |
| Recommended Storage | Refrigerator |