GA International Permanent Autoclave-Resistant Labels for Laser Printers 3"" x 2.375"" / 76.2mm x 60.3mm Rectangle White From -40C to +150C (-40F to +302F)
Numéro de catalogue. 5000572532
Permanent laser printable labels, ideal for glassware, surgical instruments, metal and plastic surfaces, trays, blister packs, and other items intended for use in steam autoclaves, ovens, and gamma irradiators. Our autoclave laser labels are durable, inert, weather & water-proof, alcohol-resistant, and have been tested to endure up to 20 autoclave cycles (at 121C/250F, 16.5 psi for 20 minutes). Compatible with all major brands of desktop laser printers.
Numéro de catalogue.
5000572532
Fournisseur
GA InternationalCode fournisseur
AKA30WH
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Permanent laser printable labels, ideal for glassware, surgical instruments, metal and plastic surfaces, trays, blister packs, and other items intended for use in steam autoclaves, ovens, and gamma irradiators. Our autoclave laser labels are durable, inert, weather & water-proof, alcohol-resistant, and have been tested to endure up to 20 autoclave cycles (at 121C/250F, 16.5 psi for 20 minutes). Compatible with all major brands of desktop laser printers.