missing translation for 'onlineSavingsMsg'
Learn More

Periodic acid, 34% w/w aq. soln.

Catalog No. AAB2017236
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2017236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2017236 Supplier Thermo Scientific Chemicals Supplier No. B2017236

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10450-60-9 | H5IO6 | 227.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10450-60-9
Molecular Formula H5IO6
Molecular Weight (g/mol) 227.94
MDL Number MFCD00011344
InChI Key TWLXDPFBEPBAQB-UHFFFAOYSA-N
Synonym orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst
PubChem CID 25289
ChEBI CHEBI:29150
IUPAC Name pentahydroxy(oxo)-$l^{7}-iodane
SMILES O[I](O)(O)(O)(O)=O

Specifications

Concentration 34% w/w aq. soln.
Sensitivity Light sensitive
Quantity 500 g
UN Number UN3098
Merck Index 14,7171
Formula Weight 227.94
Chemical Name or Material Periodic acid
Hazard Category H272-H290-H314-H335
Hazard Statement GHS H Statement
H272-H314
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P220-P221-P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING LIQUID, CORROSIVE, N.O.S.
EINECSNumber 233-937-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only