missing translation for 'onlineSavingsMsg'
Learn More

Periodic Acid, 0.5% w/v in 80% v/v Acetic Acid, Reagents

Catalog No. 01334286
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334286 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334286 Supplier Labchem Inc Supplier No. CP1217001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Periodic Acid, 0.5% w/v in 80% v/v Acetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-19-7 , 7732-18-5 , 10450-60-9
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Iodine Tetraoxide
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH <2
Color Colorless to Slight Yellow
Boiling Point 118°C
Packaging Glass Bottle
Specific Gravity 1.0447
Quantity 1 L
Physical Form Liquid
Percent Purity 0.5%
Grade Reagent
Chemical Name or Material Periodic Acid, 0.5% w/v in 80% v/v Acetic Acid