missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Perillaldehyde 90.0+%, TCI America™
$41.77 - $132.90
Chemical Identifiers
| CAS | 18031-40-8 |
|---|---|
| Molecular Formula | C10H14O |
| Molecular Weight (g/mol) | 150.221 |
| MDL Number | MFCD00001543 |
| InChI Key | RUMOYJJNUMEFDD-SNVBAGLBSA-N |
| Synonym | --perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s |
| PubChem CID | 2724159 |
| ChEBI | CHEBI:86938 |
| IUPAC Name | (4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde |
| SMILES | CC(=C)C1CCC(=CC1)C=O |
Chemical Identifiers
| 18031-40-8 | |
| 150.221 | |
| RUMOYJJNUMEFDD-SNVBAGLBSA-N | |
| 2724159 | |
| (4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde |
| C10H14O | |
| MFCD00001543 | |
| --perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s | |
| CHEBI:86938 | |
| CC(=C)C1CCC(=CC1)C=O |
Specifications
| 18031-40-8 | |
| 98°C | |
| MFCD00001543 | |
| RUMOYJJNUMEFDD-SNVBAGLBSA-N | |
| (4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde | |
| 2724159 | |
| 150.22 | |
| Liquid |
| Yellow | |
| C10H14O | |
| --perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s | |
| CC(=C)C1CCC(=CC1)C=O | |
| 150.221 | |
| CHEBI:86938 | |
| ≥90.0% (GC) | |
| (-)-Perillaldehyde |
Safety and Handling
RTECSNumber : GW2967200
TSCA : No