missing translation for 'onlineSavingsMsg'
Learn More

(-)-Perillaldehyde 90.0+%, TCI America™

Supplier:  TCI America P086625ML

Quantity 25 mL

 View more versions of this product

Catalog No. P086625ML

Only null left

Description

Description

Specifications

Chemical Identifiers

18031-40-8
150.221
RUMOYJJNUMEFDD-SNVBAGLBSA-N
2724159
(4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde
C10H14O
MFCD00001543
--perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s
CHEBI:86938
CC(=C)C1CCC(=CC1)C=O

Specifications

(-)-Perillaldehyde
Yellow
C10H14O
MFCD00001543
RUMOYJJNUMEFDD-SNVBAGLBSA-N
(4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde
2724159
150.22
Liquid
18031-40-8
98°C
25 mL
--perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s
CC(=C)C1CCC(=CC1)C=O
150.221
CHEBI:86938
≥90.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : GW2967200

TSCA : No

SDS
Documents

Documents

Promotions

Promotions