missing translation for 'onlineSavingsMsg'
Learn More

(-)-Perillaldehyde 90.0+%, TCI America™

Catalog No. P08665ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08665ML 5 mL
P086625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08665ML Supplier TCI America Supplier No. P08665ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18031-40-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00001543
InChI Key RUMOYJJNUMEFDD-SNVBAGLBSA-N
Synonym --perillaldehyde, s---perillaldehyde, s-4-prop-1-en-2-yl cyclohex-1-enecarbaldehyde, l-perillaldehyde, s-p-mentha-1,8-dien-7-al, unii-5el0y7p6lp, --perillaaldehyde, perillaldehyde,-, s-4-isopropenyl-1-cyclohexene-1-carboxaldehyde, 1-cyclohexene-1-carboxaldehyde, 4-1-methylethenyl-, 4s
PubChem CID 2724159
ChEBI CHEBI:86938
IUPAC Name (4S)-4-prop-1-en-2-ylcyclohexene-1-carbaldehyde
SMILES CC(=C)C1CCC(=CC1)C=O

Specifications

Color Yellow
Boiling Point 98°C
Quantity 5 mL
Formula Weight 150.22
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-Perillaldehyde
RTECSNumber GW2967200
TSCA No