accessibility menu, dialog, popup

UserName

H3CCH2O

CAS RN 18031-40-8

IUPAC Name: 4-(prop-1-en-2-yl)cyclohex-1-ene-1-carbaldehyde
Synonyms: (+)-perillaldehyde l-perillaldehyde perillylaldehyde 1,8-p-menthadien-7-al perillic aldehyde perillaldehyde (S)-(-)-perrillaldehyde perillyl aldehyde 1-perillaldehyde perilla aldehyde
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: RUMOYJJNUMEFDD-UHFFFAOYNA-N
SMILES: CC(=C)C1CCC(C=O)=CC1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)