Learn More
Contec™ Peridox™ Concentrate Disinfectant
Catalog No. 19165903
Change view
Click to view available options
Product Type:
Test Strips
Catalog No. Product Type
19165903 Test Strips
1 options
Catalog No. 19165903 Supplier Contec™ Supplier No. SPTS100PAA
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Peridox™ Concentrate is an EPA-registered disinfectant and cleaner that was designed to eliminate and prevent the spread of spores, as well as more than 40 other pathogens.
  • Broad spectrum disinfectant is fast acting and is effective against bacterial spores in 3 minutes, bacteria and viruses in 2 minutes, fungus in 1 minute and TB in 5 minutes.
  • Has more than 40 EPA registered claims
  • Does not contain bleach, alcohol, phenols or quaternary ammonium compounds.
  • Fast acting and will not damage stainless steel equipment
  • Compatible with all Contec mopping systems and hardware
  • Requires no rinsing and leaves no film on surfaces making it ideal for use on critical surfaces
  • With its shorter contact times and powerful wetting agents, Peridox Concentrate stays wet on the surface longer and works faster with shorter contact times and powerful wetting agents
  • Will not damage clothing, carpet, upholstery, LCD screens, and other surfaces when used as directed.
  • Patented, hyperactive chemistry provides fast results, 3-minute sporicide claim at 99.9999% efficacy
  • EPA-registered disinfectant and cleaner is ideal for use in life science cleanrooms
  • Concentrated formula provides cleanroom facilities with a cost-effective approach to use a high-level sporicidal disinfectant facility-wide
  • Compatible with most cleanroom materials of construction, and can be used on workstations, stainless steel equipment, floors, walls, and ceilings
Specifications
Product Type Test Strips
Sterility Non-sterile
For Use With (Application) Designed to eliminate and prevent the spread of spores, as well as more than 40 other pathogens