missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pericentrin Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA5115736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA5115736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA5115736 Fournisseur Invitrogen™ Code fournisseur PA5115736
Il en reste null

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Pericentrin - Centrosome Marker.

This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kD component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling.

Spécifications

Antigen Pericentrin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PCNT
Gene Accession No. O95613, P48725
Gene Alias AW476095; C86676; KEN; kendrin; KIAA0402; m239Asp; m275Asp; MOPD2; PCN; Pcnt; PCNT2; PCNTB; PCTN2; Pericentrin; pericentrin (kendrin); pericentrin 2; pericentrin-2; pericentrin-250; pericentrin-360; pericentrin-380; pericentrin-B; SCKL4
Gene Symbols PCNT
Host Species Rabbit
Immunogen A synthesized peptide derived from human PCNT(Accession O95613), corresponding to amino acid residues L3269-G3319.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18541, 309692, 5116
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats