missing translation for 'onlineSavingsMsg'
Learn More

Perfluorotriamylamine (so called), TCI America™

Catalog No. P105125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P105125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P105125G Supplier TCI America Supplier No. P105125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 338-84-1
Molecular Formula C15F33N
Molecular Weight (g/mol) 821.119
MDL Number MFCD00042367
InChI Key AQZYBQIAUSKCCS-UHFFFAOYSA-N
Synonym perfluorotripentylamine, tris undecafluoropentyl amine, perfluorotri-n-pentylamine, perfluorotriamylamine, fluorinert fc-70, perfluoro-compound fc-70, 1-pentanamine, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis undecafluoropentyl, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis 1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl pentan-1-amine, 1-pentanamine, 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-n,n-bis 1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl, fluorinert fc 70
PubChem CID 67646
IUPAC Name 1,1,2,2,3,3,4,4,5,5,5-undecafluoro-N,N-bis(1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl)pentan-1-amine
SMILES C(C(C(N(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Quantity 25 g
Formula Weight 821.12
Physical Form Liquid
Chemical Name or Material Perfluorotriamylamine (so called)
EINECSNumber (2)-3269&(2)-3525
TSCA Yes