missing translation for 'onlineSavingsMsg'
Learn More

Perfluorooctyl iodide, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 269960500

Quantity 50g
Packaging Glass bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. AC269960500


Obtenez jusqu’à 1,800 $ en produits chimiques
Afficher toutes les promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Identifiants chimiques

507-63-1
545.96
KWXGJTSJUKTDQU-UHFFFAOYSA-N
10491
C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F
C8F17I
MFCD00001064
perfluorooctyl iodide, heptadecafluoro-1-iodooctane, 1-iodoperfluorooctane, perfluoro-n-octyl iodide, 1-iodoheptadecafluorooctane, perfluoro-1-iodooctane, perfluorooctyliodide, heptadecafluoro-n-octyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodo, octane, heptadecafluoro-1-iodo
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodooctane

Spécifications

Perfluorooctyl iodide
507-63-1
160°C to 161°C
Authentic
Glass bottle
1.3286 to 1.3306
50g
2.04
Solubility in water: insoluble
C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F
545.96
545.96
99%
2.0400g/mL
>115°C
97.5% min. (GC)
C8F17I
CF3(CF2)7I
MFCD00001064
perfluorooctyl iodide, heptadecafluoro-1-iodooctane, 1-iodoperfluorooctane, perfluoro-n-octyl iodide, 1-iodoheptadecafluorooctane, perfluoro-1-iodooctane, perfluorooctyliodide, heptadecafluoro-n-octyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodo, octane, heptadecafluoro-1-iodo
KWXGJTSJUKTDQU-UHFFFAOYSA-N
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodooctane
10491
98%
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-079-5

RTECSNumber : RG9700050

TSCA : TSCA

FDS
Documents

Documents

Promotions

Promotions

Promo
Date d’expiration: 06-30-2027
Obtenez jusqu’à 1,800 $ en produits chimiques
Dépensez 250 $ ou plus en produits chimiques Thermo Fisher Scientific et obtenez jusqu’à 1 800 $ en produits chimiques.

Ajout...