missing translation for 'onlineSavingsMsg'
Learn More

Perfex™ Broom and Brush Handles

Catalog No. 19042041
Change view
Click to view available options
Product Type:
Adjustable Polymer Handle
Hardwood Handle
Length (English):
34 to 62 in. with Long Connector
34 to 62 in. with Short Connector
60 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Length (English)
19042041 Adjustable Polymer Handle 34 to 62 in. with Short Connector
19042040 Adjustable Polymer Handle 34 to 62 in. with Long Connector
19042037 Hardwood Handle 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19042041 Supplier Perfex™ Supplier No. 8800S

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hardwood Handles.
The 60 in. (152cm) hardwood handle should be used with Model Nos. 2618, 2624, 2518, 2524, 2418 and 3010, while the 48 in. (122cm) hardwood handle is recommended for Model Nos. 2200 and 2300.
Polymer Adjustable Handles.
The Polymer Adjustable Handle adjusts in length from 34 to 62 in. (86 to 157cm). With the short connector end, use with Model Nos. 2200 and 2300. With a long connector end, it is recommended for use with Perfex Model Nos. 2618, 2624, 2518, 2524, 2418 and 3010, as well as squeegee Model Nos. 9518, 9524, and 9530.

Specifications

Product Type Adjustable Polymer Handle
Color Red, White, Blue
For Use With (Application) Reduces strain and transfers maximum energy with minimum exertion
For Use With (Equipment) Perfex Model Nos. 2618, 2624, 2518, 2524, 2418 and 3010, as well as squeegee Model Nos. 9518, 9524 and 9530
Length (English) 34 to 62 in. with Short Connector
Length (Metric) 86 to 157 cm
Material Polyester