missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, Ultratrace, 65-71%, Spectrum™ Chemical

Catalog No. 18612189
Change view
Click to view available options
Quantity:
500 mL
Packaging:
PTFE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612189 500 mL PTFE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18612189 Supplier Spectrum Chemical Mfg Cor Supplier No. P1357500MLTF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

CAS Max % 1
Packaging PTFE Bottle
Quantity 500 mL
Percent Purity 65 to 71%
Grade High Purity Solvent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.