missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid Solution, 1.0M, Honeywell™
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Spécifications
Spécifications
| Quantity | 1 L |
| Linear Formula | HClO4 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 100.46g/mol |
| Chemical Name or Material | Perchloric acid solution |
Sécurité et manipulation
| Hazard Statement | P280-P305 + P351 + P338-P337 + P313 |