missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, 0.0200 N (N/50) in Glacial Acetic Acid, Ricca Chemical

Catalog No. 551432
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
551432 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 551432 Supplier Ricca Chemical Company Supplier No. 551432

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64-19-7 , 108-24-7 , 7601-90-3 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless to Yellow
Concentration 0.02Normal
CAS Min % 96.0, 2.07, 0.26
CAS Max % 100.0, 2.15, 0.27
Packaging Amber Glass Bottle
Solvent or Matrix Glacial Acetic Acid
Quantity 1 L
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Perchloric Acid