CAS RN 108-24-7
IUPAC Name:
acetyl acetate
Synonyms:
acetic anhydride acetic anhydride, 3H-labeled octowy bezwodnik acetic anhydride (substance) 3h-labeled acetic anhydride 1,1'-anhydride acetic acid ethanoic anhydrate anhydrid kyseliny octove anhydride acetique azijnzuuranhydride
Molecular Weight (g/mol):
102.09
Molecular Formula:
C4H6O3
InChi Key:
WFDIJRYMOXRFFG-UHFFFAOYSA-N
SMILES:
CC(=O)OC(C)=O
1
–
1
of
1
results
1
Acetic Anhydride (Certified ACS), Fisher Chemical™
CAS: 108-24-7 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.089 MDL Number: MFCD00008705 InChI Key: WFDIJRYMOXRFFG-UHFFFAOYSA-N Synonym: acetic anhydride,acetanhydride,acetic acid, anhydride,acetic oxide,acetyl oxide,ethanoic anhydride,acetyl ether,acetyl anhydride,acetic acid anhydride,anhydride acetique PubChem CID: 7918 ChEBI: CHEBI:36610 IUPAC Name: acetyl acetate SMILES: CC(=O)OC(=O)C