missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PeproGMP® Human Activin A Recombinant Protein, PeproTech®

Numéro de catalogue. 01672638
Encompass
Modifier l'affichage
Click to view available options
Quantité:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
01672638 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01672638 Fournisseur Gibco™ Code fournisseur GMP12014E50UG

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Recombinant Protein

Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.

Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.

Spécifications

Numéro d'enregistrement P08476
À utiliser avec (application) Control, Western Blot Control, Bioactivity
Format Lyophilized
Formule Protein with no preservative
ID du gène (Entrez) 3624
Poids moléculaire (g/mol) 26 kDa
Nom PeproGMP™ Human Activin A
Méthode de purification Purified
Quantité 50 μg
Conditions de stockage -20°C or -80°C if preferred
Statut réglementaire GMP
Concentration en endotoxines ≤0.1 EU/μg
Alias du gène activin A; activin beta-A chain; BMP7; BMP-7; Bone morphogenetic protein 7; EDF; Eptotermin alfa; erythroid differentiation factor; erythroid differentiation protein; follicle-stimulating hormone-releasing protein; FRP; FSH-releasing protein; INHB; Inhba; inhibin B subunit; inhibin beta A chain; inhibin beta A protein; inhibin beta A subunit; inhibin beta-A; inhibin beta-A-subunit; inhibin beta-subunit; inhibin subunit beta A; inhibin, beta A; inhibin, beta A (activin A, activin AB alpha polypeptide); Inhibin, beta-1; OP1; OP-1; Osteogenic protein 1; unnamed protein product
Nom usuel Activin A
Symbole de gène Inhba
Activité biologique Determined by the proliferation of mouse MPC-11 cells. PeproGMP Recombinant Human Activin A has been tested against the WHO standard NIBSC 91/626.
Type de produit Protein
Conjugué Unconjugated
Recombinant Recombinant
Séquence GLECDGKVNI CCKKQFFVSF KDIGWNDWII APSGYHANYC EGECPSHIAG TSGSSLSFHS TVINHYRMRG HSPFANLKSC CVPTKLRPMS MLYYDDGQNI IKKDIQNMIV EECGCS
Contenu et stockage -20°Cor to +80°C if preferred
Activité Determined by the proliferation of mouse MPC-11 cells. PeproGMP Recombinant Human Activin A has been tested against the WHO standard NIBSC 91/626. Find the International Unit (IU) value <a href='https://www.thermofisher.com/us/en/home/life-science/cell-culture/cell-culture-learning-center/recombinant-protein-information/international-units.html'>here</a>.
Système d’expression E. coli
Forme Lyophilized
Grade de qualité ou de pureté ≥ 98% by SDS-PAGE gel and HPLC analyses.
Afficher plus de résultats Afficher moins de résultats