missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PeproGMP® Human EGF Recombinant Protein, PeproTech®

Numéro de catalogue. 01672630
Encompass
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
01672630 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01672630 Fournisseur Gibco™ Code fournisseur GMP10015100UG

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Recombinant Protein

Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.

EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.

Spécifications

Numéro d'enregistrement P01133
À utiliser avec (application) Control, Western Blot Control, Bioactivity
Format Lyophilized
Formule Protein with no preservative
ID du gène (Entrez) 1950
Poids moléculaire (g/mol) 6.2 kDa
Nom PeproGMP™ Human EGF
Méthode de purification Purified
Quantité 100 μg
Conditions de stockage -20°C or -80°C if preferred
Statut réglementaire GMP
Concentration en endotoxines ≤0.1 EU/μg
Alias du gène AI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; Urogastrone
Nom usuel EGF
Symbole de gène EGF
Activité biologique Determined by the proliferation of mouse Balb/c 3T3 cells. PeproGMP Recombinant Human EGF has been tested against the WHO standard NIBSC 91/530.
Type de produit Protein
Conjugué Unconjugated
Recombinant Recombinant
Séquence NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR
Contenu et stockage -20°Cor to +80°C if preferred
Activité Determined by the proliferation of mouse Balb/c 3T3 cells. PeproGMP Recombinant Human EGF has been tested against the WHO standard NIBSC 91/530. Find the International Unit (IU) value <a href='https://www.thermofisher.com/us/en/home/life-science/cell-culture/cell-culture-learning-center/recombinant-protein-information/international-units.html'>here</a>.
Système d’expression E. coli
Forme Lyophilized
Grade de qualité ou de pureté ≥ 98% by SDS-PAGE gel and HPLC analyses.
Afficher plus de résultats Afficher moins de résultats