missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentyl Valerate 98.0+%, TCI America™
$48.46 - $328.08
Chemical Identifiers
| CAS | 2173-56-0 |
|---|---|
| Molecular Formula | C10H20O2 |
| Molecular Weight (g/mol) | 172.27 |
| MDL Number | MFCD00042904 |
| InChI Key | FGPPDYNPZTUNIU-UHFFFAOYSA-N |
| Synonym | pentyl valerate, amyl valerate, pentanoic acid, pentyl ester, n-pentyl valerate, amyl valerianate, valeric acid, pentyl ester, 1-pentyl n-valerate, valeric acid pentyl ester, pentanoicacid, pentyl ester, pentanoic acid pentyl ester |
| PubChem CID | 62433 |
| IUPAC Name | pentyl pentanoate |
| SMILES | CCCCCOC(=O)CCCC |
Chemical Identifiers
| 2173-56-0 | |
| 172.27 | |
| FGPPDYNPZTUNIU-UHFFFAOYSA-N | |
| 62433 | |
| CCCCCOC(=O)CCCC |
| C10H20O2 | |
| MFCD00042904 | |
| pentyl valerate, amyl valerate, pentanoic acid, pentyl ester, n-pentyl valerate, amyl valerianate, valeric acid, pentyl ester, 1-pentyl n-valerate, valeric acid pentyl ester, pentanoicacid, pentyl ester, pentanoic acid pentyl ester | |
| pentyl pentanoate |
Specifications
| 2173-56-0 | |
| 205°C | |
| MFCD00042904 | |
| pentyl valerate, amyl valerate, pentanoic acid, pentyl ester, n-pentyl valerate, amyl valerianate, valeric acid, pentyl ester, 1-pentyl n-valerate, valeric acid pentyl ester, pentanoicacid, pentyl ester, pentanoic acid pentyl ester | |
| CCCCCOC(=O)CCCC | |
| 172.27 | |
| 172.27 | |
| Liquid |
| Colorless | |
| C10H20O2 | |
| 25 mL | |
| FGPPDYNPZTUNIU-UHFFFAOYSA-N | |
| pentyl pentanoate | |
| 62433 | |
| ≥98.0% (GC) | |
| Pentyl Valerate |
Safety and Handling
EINECSNumber : (2)-0776
RTECSNumber : SA4250000
TSCA : Yes