missing translation for 'onlineSavingsMsg'
Learn More

Pentyl Valerate 98.0+%, TCI America™

Supplier:  TCI America P192925ML

 View more versions of this product

Catalog No. P192925ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pentyl Valerate
Colorless
C10H20O2
25 mL
FGPPDYNPZTUNIU-UHFFFAOYSA-N
pentyl pentanoate
62433
≥98.0% (GC)
2173-56-0
205°C
MFCD00042904
pentyl valerate, amyl valerate, pentanoic acid, pentyl ester, n-pentyl valerate, amyl valerianate, valeric acid, pentyl ester, 1-pentyl n-valerate, valeric acid pentyl ester, pentanoicacid, pentyl ester, pentanoic acid pentyl ester
CCCCCOC(=O)CCCC
172.27
172.27
Liquid

Chemical Identifiers

2173-56-0
172.27
FGPPDYNPZTUNIU-UHFFFAOYSA-N
62433
CCCCCOC(=O)CCCC
C10H20O2
MFCD00042904
pentyl valerate, amyl valerate, pentanoic acid, pentyl ester, n-pentyl valerate, amyl valerianate, valeric acid, pentyl ester, 1-pentyl n-valerate, valeric acid pentyl ester, pentanoicacid, pentyl ester, pentanoic acid pentyl ester
pentyl pentanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0776

RTECSNumber : SA4250000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions