accessibility menu, dialog, popup

UserName

CAS RN 2173-56-0

H3COOCH3

CAS RN 2173-56-0

IUPAC Name: pentyl pentanoate
Synonyms: pentyl pentanoate
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: FGPPDYNPZTUNIU-UHFFFAOYSA-N
SMILES: CCCCCOC(=O)CCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results