missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentyl Valerate 98.0+%, TCI America™
Supplier: TCI America P1929500ML
Specifications
| Pentyl Valerate | |
| Colorless | |
| C10H20O2 | |
| 500 mL | |
| FGPPDYNPZTUNIU-UHFFFAOYSA-N | |
| pentyl pentanoate | |
| 62433 | |
| ≥98.0% (GC) |
| 2173-56-0 | |
| 205°C | |
| MFCD00042904 | |
| pentyl valerate, amyl valerate, pentanoic acid, pentyl ester, n-pentyl valerate, amyl valerianate, valeric acid, pentyl ester, 1-pentyl n-valerate, valeric acid pentyl ester, pentanoicacid, pentyl ester, pentanoic acid pentyl ester | |
| CCCCCOC(=O)CCCC | |
| 172.27 | |
| 172.27 | |
| Liquid |
Chemical Identifiers
| 2173-56-0 | |
| 172.27 | |
| FGPPDYNPZTUNIU-UHFFFAOYSA-N | |
| 62433 | |
| CCCCCOC(=O)CCCC |
| C10H20O2 | |
| MFCD00042904 | |
| pentyl valerate, amyl valerate, pentanoic acid, pentyl ester, n-pentyl valerate, amyl valerianate, valeric acid, pentyl ester, 1-pentyl n-valerate, valeric acid pentyl ester, pentanoicacid, pentyl ester, pentanoic acid pentyl ester | |
| pentyl pentanoate |
Safety and Handling
EINECSNumber : (2)-0776
RTECSNumber : SA4250000
TSCA : Yes