missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentyl isocyanate, 98%
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | Pentyl isocyanate |
| Nom Remarque | 98% |
| Couleur | Incolore |
| Densité | 0.8700g/mL |
| Point d’ébullition | 136,0 °C à 137,0 °C |
| Point d’éclair | 32 °C |
| Spectre infrarouge | Authentique |
| Fourchette de pourcentages d’essai | 97,5% min. (GC) |
| Emballage | Bouteille en verre |
| Indice de réfraction | 1,4130 à 1,4150 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes skin irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if inhaled. May cause respiratory irritation. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. |
RUO – Usage réservé à la recherche