missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentane 97.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 500 mL |
| Physical Form | Clear Liquid at 20°C |
| UN Number | 1265 |
| Boiling Point | 36°C |
| Formula Weight | 72.15 |
| Percent Purity | ≥97.0% (GC) |
| Chemical Name or Material | Pentane |
Safety and Handling
| EINECSNumber | (2)-0005 |
| RTECSNumber | RZ9450000 |
| TSCA | Yes |