missing translation for 'onlineSavingsMsg'
Learn More

Pentane 97.0+%, TCI America™

Numéro de catalogue. P2621100ML
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P2621100ML 100 mL
P2621500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P2621100ML Fournisseur TCI America Code fournisseur P2621100ML
Il en reste null

Identifiants chimiques

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Spécifications

Color Colorless
Quantity 100 mL
Physical Form Clear Liquid at 20°C
UN Number 1265
Boiling Point 36°C
Formula Weight 72.15
Percent Purity ≥97.0% (GC)
Chemical Name or Material Pentane
EINECSNumber (2)-0005
RTECSNumber RZ9450000
TSCA Yes