missing translation for 'onlineSavingsMsg'
Learn More

Pentane 99.5+%, TCI America™

Numéro de catalogue. S02775ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
S02775ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. S02775ML Fournisseur TCI America Code fournisseur S02775ML
Il en reste null

Identifiants chimiques

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Spécifications

Color Colorless
Quantity 5 mL
Physical Form Clear Liquid at 20°C
UN Number 1265
Boiling Point 36°C
Formula Weight 72.15
Percent Purity ≥99.5% (GC)
Chemical Name or Material Pentane [Standard Material for GC]
EINECSNumber (2)-0005
RTECSNumber RZ9450000
TSCA Yes