missing translation for 'onlineSavingsMsg'
Learn More

Pentane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9%, Honeywell Burdick & Jackson

Catalog No. 6001126
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001126 6 x 1 L Glass Bottle
6001124 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001126 Supplier Honeywell Chemicals Supplier No. 3121L
Only null left

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

CAS Max % 100.0000%
Density 0.626 g/cm3
Quantity 6 x 1 L
Flash Point −49°C
Packaging Glass Bottle
UN Number UN1265
Beilstein 969132
Boiling Point 36°C
Formula Weight 72.17
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Pentane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Pentanes