missing translation for 'onlineSavingsMsg'
Learn More

Pentane, 98% min., OmniSolv™, MilliporeSigma™

Catalog No. MPX01671
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPX01671 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX01671 Supplier Technidata Supplier No. PX01671

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 109-66-0
Color Colorless
Density 0.63g/cm3 (20°C)
Quantity 4 L
Explosion Limit 1.4 to 8%(V)
Ignition Point 285°C
Physical Form Liquid
Assay Percent Range 98% min.
Molecular Formula C5H12
Linear Formula CH3(CH2)3CH3
Vapor Pressure 570 hPa (20°C)
MDL Number MFCD00009498
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILES CCCCC
IUPAC Name pentane
Molecular Weight (g/mol) 72.15
PubChem CID 8003
ChEBI CHEBI:37830
Formula Weight 72.15
Chemical Name or Material Pentane
Show More Show Less
Recommended Storage Store at +15°C to +25°C