missing translation for 'onlineSavingsMsg'
Learn More

(Pentamethylcyclopentadienyl)rhodium(III) Dichloride Dimer 96.0+%, TCI America™

Numéro de catalogue. P1788200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P1788200MG 200 mg
P17881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P1788200MG Fournisseur TCI America Code fournisseur P1788200MG
Il en reste null

Identifiants chimiques

CAS 12354-85-7
Molecular Formula C20H30Cl4Rh2
Molecular Weight (g/mol) 618.07
MDL Number MFCD00061552
InChI Key QNIVKTTWBMFSBR-UHFFFAOYSA-J
Synonym Dichloro(pentamethylcyclopentadienyl)rhodium(III) Dimer
PubChem CID 74222901
IUPAC Name 1,2,3,4,5-pentamethylcyclopenta-1,3-diene;rhodium(2+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Rh++].[Rh++].C[C]1C(C)=C(C)C(C)=C1C.C[C]1C(C)=C(C)C(C)=C1C

Spécifications

Quantity 200 mg
Formula Weight 618.07
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (Pentamethylcyclopentadienyl)rhodium(III) Dichloride Dimer