missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorophenyl Methacrylate (stabilized with MEHQ) 97.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 252.14 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Pentafluorophenyl Methacrylate (stabilized with MEHQ) |
Sécurité et manipulation
| TSCA | No |
| Recommended Storage | Refrigerator |