missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 4-(1,3-oxazol-5-yl)benzoate, 97%, Thermo Scientific™

Numéro de catalogue. CC14526DA
Modifier l'affichage
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
CC14526DA Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC14526DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC14526DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Spécifications

Nom ou substance chimique Pentafluorophenyl 4-(1, 3-oxazol-5-yl)benzoate
CAS 898288-91-0
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Formule moléculaire C16H6F5NO3
Quantité 1g
Numéro MDL MFCD09025822
Synonyme pentafluorophenyl 4-1,3-oxazol-5-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-1,3-oxazol-5-yl benzoate, perfluorophenyl 4-oxazol-5-yl benzoate, 4-5-oxazolyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester, benzoic acid,4-5-oxazolyl-, 2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentakis fluoranyl phenyl 4-1,3-oxazol-5-yl benzoate
Clé InChI FWUPBFHGEDUEID-UHFFFAOYSA-N
SOURIRES C1=CC(=CC=C1C2=CN=CO2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F
Nom de l’IUPAC (2,3,4,5,6-pentafluorophenyl) 4-(1,3-oxazol-5-yl)benzoate
Poids moléculaire (g/mol) 355.22
PubChem CID 18525706
Poids de formule 355.22
Pourcentage de pureté 97%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.