missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 2-thiomorpholin-4-ylisonicotinate, 97%, Thermo Scientific™

Numéro de catalogue. CC50526DA
Modifier l'affichage
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
CC50526DA Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC50526DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC50526DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Spécifications

Nom ou substance chimique Pentafluorophenyl 2-thiomorpholin-4-ylisonicotinate
CAS 934570-42-0
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Formule moléculaire C16H11F5N2O2S
Quantité 1g
Numéro MDL MFCD09702363
Synonyme pentafluorophenyl 2-thiomorpholin-4-ylisonicotinate, pentafluorophenyl 2-thiomorpholin-4-ylpyridine-4-carboxylate, pentafluorophenyl 2-thiomorpholin-4-yl pyridine-4-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-thiomorpholin-4-ylpyridine-4-carboxylate, 2,3,4,5,6-pentafluorophenyl 2-thiomorpholin-4-yl pyridine-4-carboxylate, 2-thiomorpholin-4-yl-4-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
Clé InChI CXWZKWSFJLNFMF-UHFFFAOYSA-N
SOURIRES FC1=C(F)C(F)=C(OC(=O)C2=CC(=NC=C2)N2CCSCC2)C(F)=C1F
Nom de l’IUPAC (2,3,4,5,6-pentafluorophenyl) 2-thiomorpholin-4-ylpyridine-4-carboxylate
Poids moléculaire (g/mol) 390.33
PubChem CID 24229510
Poids de formule 390.33
Pourcentage de pureté 97%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.