missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 2-morpholino-1,3-thiazole-4-carboxylate, 97%, Thermo Scientific™

Numéro de catalogue. p-3541154
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC62826CB 250mg Amber Glass Bottle
CC62826DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC62826CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC62826CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 921939-02-8
Formule moléculaire C14H9F5N2O3S
Poids moléculaire (g/mol) 380.29
Numéro MDL MFCD09817538
Clé InChI NABJYXXXUIZUOT-UHFFFAOYSA-N
Synonyme pentafluorophenyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate, pentafluorophenyl 2-morpholino-1,3-thiazole-4-carboxylate, 2,3,4,5,6-pentafluorophenyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate, 2-4-morpholinyl-4-thiazolecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229699
Nom de l’IUPAC (2,3,4,5,6-pentafluorophenyl) 2-morpholin-4-yl-1,3-thiazole-4-carboxylate
SOURIRES FC1=C(F)C(F)=C(OC(=O)C2=CSC(=N2)N2CCOCC2)C(F)=C1F

Spécifications

Nom ou substance chimique Pentafluorophenyl 2-morpholino-1, 3-thiazole-4-carboxylate
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 380.29
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.