missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenol 98.0+%, TCI America™

Catalog No. P091925G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P091925G 25 g
P091910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P091925G Supplier TCI America Supplier No. P091925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 771-61-9
Molecular Formula C6HF5O
Molecular Weight (g/mol) 184.07
MDL Number MFCD00002156
InChI Key XBNGYFFABRKICK-UHFFFAOYSA-N
Synonym pentafluorophenol, phenol, pentafluoro, perfluorophenol, unii-a2ycf0yuha, a2ycf0yuha, 2,3,4,5,6-pentafluorohydroxybenzene, c6f5oh, 2,3,4,5,6-pentafluoro-phenol, phenol, 2,3,4,5,6-pentafluoro, hopfp
PubChem CID 13041
IUPAC Name pentafluorophenol
SMILES OC1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Color White-Yellow
Boiling Point 143°C
Quantity 25 g
Formula Weight 184.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Pentafluorophenol
RTECSNumber SM6680000
TSCA Yes