missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzyl Alcohol 96.0+%, TCI America™

Catalog No. P08495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08495G 5 g
P084925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08495G Supplier TCI America Supplier No. P08495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 440-60-8
Molecular Formula C7H3F5O
Molecular Weight (g/mol) 198.092
MDL Number MFCD00004602
InChI Key PGJYYCIOYBZTPU-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzyl alcohol, pentafluorobenzyl alcohol, perfluorophenyl methanol, pentafluorophenyl methanol, 2,3,4,5,6-pentafluorophenyl methanol, 2,3,4,5,6-pentafluorobenzenemethanol, hydroxymethyl pentafluorobenzene, benzenemethanol, 2,3,4,5,6-pentafluoro, pentafluorobenzylalcohol, 2,3,4,5,6-pentafluorobenzylalcohol
PubChem CID 9923
ChEBI CHEBI:44903
IUPAC Name (2,3,4,5,6-pentafluorophenyl)methanol
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)O

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 182°C
Quantity 5 g
Formula Weight 198.09
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Pentafluorobenzyl Alcohol
RTECSNumber DP0695000
TSCA No