missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorobenzene 98.0+%, TCI America™

Catalog No. P09235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09235G 5 g
P092325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09235G Supplier TCI America Supplier No. P09235G

Chemical Identifiers

CAS 363-72-4
Molecular Formula C6HF5
Molecular Weight (g/mol) 168.07
MDL Number MFCD00000286
InChI Key WACNXHCZHTVBJM-UHFFFAOYSA-N
Synonym pentafluorobenzene, benzene, pentafluoro, unii-ih36lu53xs, benzene, 1,2,3,4,5-pentafluoro, 2,3,4,5,6-pentafluorobenzene, ih36lu53xs, 1,2,3,4,5-pentafluoro-benzene, pentafluorophenyl, benzenepentafluoride, pubchem2310
PubChem CID 9696
IUPAC Name 1,2,3,4,5-pentafluorobenzene
SMILES FC1=CC(F)=C(F)C(F)=C1F

Specifications

Melting Point -48°C
Color Yellow
Boiling Point 85°C
Quantity 5 g
UN Number 1993
Formula Weight 168.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluorobenzene
EINECSNumber (3)-4337
RTECSNumber DA6651600
TSCA Yes