missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentadecanoic Acid 98.0+%, TCI America™
Chemical Identifiers
| CAS | 1002-84-2 |
|---|---|
| Molecular Formula | C15H30O2 |
| Molecular Weight (g/mol) | 242.403 |
| MDL Number | MFCD00002745 |
| InChI Key | WQEPLUUGTLDZJY-UHFFFAOYSA-N |
| Synonym | pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure |
| PubChem CID | 13849 |
| ChEBI | CHEBI:42504 |
| IUPAC Name | pentadecanoic acid |
| SMILES | CCCCCCCCCCCCCCC(=O)O |
Chemical Identifiers
| C15H30O2 | |
| MFCD00002745 | |
| pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure | |
| CHEBI:42504 | |
| CCCCCCCCCCCCCCC(=O)O |
Specifications
| 1002-84-2 | |
| White | |
| C15H30O2 | |
| pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure | |
| CCCCCCCCCCCCCCC(=O)O | |
| 242.403 | |
| CHEBI:42504 | |
| ≥98.0% (GC,T) | |
| Pentadecanoic Acid |
| 53°C | |
| 339°C | |
| MFCD00002745 | |
| WQEPLUUGTLDZJY-UHFFFAOYSA-N | |
| pentadecanoic acid | |
| 13849 | |
| 242.40 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : RZ1925000
TSCA : Yes