missing translation for 'onlineSavingsMsg'
Learn More

Pentadecanoic Acid 98.0+%, TCI America™

Catalog No. P003525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P003525G 25 g
P0035100G 100 g
P0035500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P003525G Supplier TCI America Supplier No. P003525G

Chemical Identifiers

CAS 1002-84-2
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.403
MDL Number MFCD00002745
InChI Key WQEPLUUGTLDZJY-UHFFFAOYSA-N
Synonym pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure
PubChem CID 13849
ChEBI CHEBI:42504
IUPAC Name pentadecanoic acid
SMILES CCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 53°C
Color White
Boiling Point 339°C
Quantity 25 g
Formula Weight 242.40
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pentadecanoic Acid
EINECSNumber (2)-0608
RTECSNumber RZ1925000
TSCA Yes