missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentadecanoic Acid 98.0+%, TCI America™
Supplier: TCI America P0035100G
| Quantity | 100 g |
|---|
Chemical Identifiers
| C15H30O2 | |
| MFCD00002745 | |
| pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure | |
| CHEBI:42504 | |
| CCCCCCCCCCCCCCC(=O)O |
Specifications
| Pentadecanoic Acid | |
| 53°C | |
| 339°C | |
| 100 g | |
| pentadecylic acid, n-pentadecanoic acid, pentadecyclic acid, n-pentadecylic acid, unii-ccw02d961f, pentadecyclate, n-pentadecylate, n-pentadecanoate, n-pentadecanoicacid, pentadecansäure | |
| CCCCCCCCCCCCCCC(=O)O | |
| 242.403 | |
| CHEBI:42504 | |
| ≥98.0% (GC,T) |
| 1002-84-2 | |
| White | |
| C15H30O2 | |
| MFCD00002745 | |
| WQEPLUUGTLDZJY-UHFFFAOYSA-N | |
| pentadecanoic acid | |
| 13849 | |
| 242.40 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : RZ1925000
TSCA : Yes