accessibility menu, dialog, popup

UserName

H3COHO

CAS RN 1002-84-2

IUPAC Name: pentadecanoic acid
Synonyms: pentadecanoic acid pentadecyclate pentadecyclic acid
Molecular Weight (g/mol): 242.4
Molecular Formula: C15H30O2
InChi Key: WQEPLUUGTLDZJY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC(O)=O
Boiling Point 
  • (3)
  • (8)
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (8)
  • (2)
  • (3)
  • (6)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)