missing translation for 'onlineSavingsMsg'
Learn More

Pentachlorothioanisole 98.0+%, TCI America™

Numéro de catalogue. P10671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P10671G 1 g
P10675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P10671G Fournisseur TCI America Code fournisseur P10671G
Il en reste null

Identifiants chimiques

CAS 1825-19-0
Molecular Formula C7H3Cl5S
Molecular Weight (g/mol) 296.41
MDL Number MFCD00013622
InChI Key LGZZJTIUEJNNKV-UHFFFAOYSA-N
Synonym Methyl Pentachlorophenyl Sulfide, 1,2,3,5,6-Pentachloro-4-(methylthio)benzene
PubChem CID 15766
IUPAC Name 1,2,3,4,5-pentachloro-6-(methylsulfanyl)benzene
SMILES CSC1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl

Spécifications

Melting Point 95°C
Color White-Yellow
Quantity 1 g
Formula Weight 296.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentachlorothioanisole
RTECSNumber WQ5520000
TSCA No