missing translation for 'onlineSavingsMsg'
Learn More

Pendimethalin 98.0+%, TCI America™

Numéro de catalogue. P19645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P19645G 5 g
P196425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P19645G Fournisseur TCI America Code fournisseur P19645G
Il en reste null

Identifiants chimiques

CAS 40487-42-1
Molecular Formula C13H19N3O4
Molecular Weight (g/mol) 281.312
MDL Number MFCD00055332
InChI Key CHIFOSRWCNZCFN-UHFFFAOYSA-N
Synonym pendimethalin, pendimethaline, penoxaline, prowl, herbadox, penoxalin, phenoxalin, accotab, stomp, way up
PubChem CID 38479
ChEBI CHEBI:83569
IUPAC Name 3,4-dimethyl-2,6-dinitro-N-pentan-3-ylaniline
SMILES CCC(CC)NC1=C(C=C(C(=C1[N+](=O)[O-])C)C)[N+](=O)[O-]

Spécifications

Melting Point 58°C
Quantity 5 g
UN Number 3077
Formula Weight 281.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pendimethalin
RTECSNumber BX5470000
TSCA No