missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Penciclovir

$244.94 - $715.00

Chemical Identifiers

CAS 39809-25-1
Molecular Formula C10H15N5O3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC468730010
View Documents
Thermo Scientific Chemicals
468730010
1 g
Each for $715.00
Only null left
 
AC468732500
View Documents
Thermo Scientific Chemicals
468732500
250 mg
Each for $244.94
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39809-25-1
C10H15N5O3

Specifications

39809-25-1
Solubility aq. soln.: 1.7 mg/mL (20°C).
C10H15N5O3
JNTOCHDNEULJHD-UHFFFAOYSA-N
2-amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-6,9-dihydro-3H-purin-6-one
Penciclovir
1 g
275.0°C to 277.0°C
253.26
NC1=NC(=O)C2=C(N1)N(CCC(CO)CO)C=N2
253.26
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes serious eye irritation.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only