missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Penciclovir

Supplier:  Thermo Scientific Chemicals 468730010

 View more versions of this product

Catalog No. AC468730010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Penciclovir
1 g
275.0°C to 277.0°C
253.26
NC1=NC(=O)C2=C(N1)N(CCC(CO)CO)C=N2
253.26
39809-25-1
Solubility aq. soln.: 1.7 mg/mL (20°C).
C10H15N5O3
JNTOCHDNEULJHD-UHFFFAOYSA-N
2-amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-6,9-dihydro-3H-purin-6-one

Chemical Identifiers

39809-25-1
C10H15N5O3
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes serious eye irritation.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only