missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pemetrexed Hydrate 98.0+%, TCI America™
$149.13 - $372.81
Chemical Identifiers
| CAS | 137281-23-3 |
|---|---|
| Molecular Formula | C20H21N5O6 |
| Molecular Weight (g/mol) | 427.417 |
| MDL Number | MFCD19105065 |
| InChI Key | WBXPDJSOTKVWSJ-ZDUSSCGKSA-N |
| Synonym | N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid |
| PubChem CID | 446556 |
| ChEBI | CHEBI:63616 |
| IUPAC Name | (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid |
| SMILES | C1=CC(=CC=C1CCC2=CNC3=C2C(=O)N=C(N3)N)C(=O)NC(CCC(=O)O)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P2288250MG
|
TCI America
P2288250MG |
250 mg |
Each for $149.13
|
|
|||||
|
P22881G
|
TCI America
P22881G |
1 g |
Each for $372.81
|
|
|||||
Chemical Identifiers
| 137281-23-3 | |
| 427.417 | |
| WBXPDJSOTKVWSJ-ZDUSSCGKSA-N | |
| 446556 | |
| (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid |
| C20H21N5O6 | |
| MFCD19105065 | |
| N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid | |
| CHEBI:63616 | |
| C1=CC(=CC=C1CCC2=CNC3=C2C(=O)N=C(N3)N)C(=O)NC(CCC(=O)O)C(=O)O |
Specifications
| 137281-23-3 | |
| C20H21N5O6 | |
| Blue/White | |
| WBXPDJSOTKVWSJ-ZDUSSCGKSA-N | |
| (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid | |
| 250 mg | |
| CHEBI:63616 | |
| ≥98.0% (HPLC,T) |
| Powder/Crystals | |
| MFCD19105065 | |
| N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid | |
| C1=CC(=CC=C1CCC2=CNC3=C2C(=O)N=C(N3)N)C(=O)NC(CCC(=O)O)C(=O)O | |
| 427.417 | |
| 446556 | |
| 427.42 | |
| Pemetrexed Hydrate |
Safety and Handling
RTECSNumber : LZ9950100
TSCA : No
Recommended Storage : Refrigerator