missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pemetrexed Hydrate 98.0+%, TCI America™
Supplier: TCI America P2288250MG
Specifications
| Pemetrexed Hydrate | |
| Powder/Crystals | |
| MFCD19105065 | |
| N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid | |
| C1=CC(=CC=C1CCC2=CNC3=C2C(=O)N=C(N3)N)C(=O)NC(CCC(=O)O)C(=O)O | |
| 427.417 | |
| 446556 | |
| 427.42 |
| 137281-23-3 | |
| C20H21N5O6 | |
| Blue/White | |
| WBXPDJSOTKVWSJ-ZDUSSCGKSA-N | |
| (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid | |
| 250 mg | |
| CHEBI:63616 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 137281-23-3 | |
| 427.417 | |
| WBXPDJSOTKVWSJ-ZDUSSCGKSA-N | |
| 446556 | |
| (2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid |
| C20H21N5O6 | |
| MFCD19105065 | |
| N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid | |
| CHEBI:63616 | |
| C1=CC(=CC=C1CCC2=CNC3=C2C(=O)N=C(N3)N)C(=O)NC(CCC(=O)O)C(=O)O |
Safety and Handling
RTECSNumber : LZ9950100
TSCA : No
Recommended Storage : Refrigerator