missing translation for 'onlineSavingsMsg'
Learn More

Pemetrexed Hydrate 98.0+%, TCI America™

Supplier:  TCI America P22881G

 View more versions of this product

Catalog No. P22881G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pemetrexed Hydrate
Powder/Crystals
MFCD19105065
N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid
C1=CC(=CC=C1CCC2=CNC3=C2C(=O)N=C(N3)N)C(=O)NC(CCC(=O)O)C(=O)O
427.417
446556
427.42
137281-23-3
C20H21N5O6
Blue/White
WBXPDJSOTKVWSJ-ZDUSSCGKSA-N
(2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid
1 g
CHEBI:63616
≥98.0% (HPLC,T)

Chemical Identifiers

137281-23-3
427.417
WBXPDJSOTKVWSJ-ZDUSSCGKSA-N
446556
(2S)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioic acid
C20H21N5O6
MFCD19105065
N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid
CHEBI:63616
C1=CC(=CC=C1CCC2=CNC3=C2C(=O)N=C(N3)N)C(=O)NC(CCC(=O)O)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : LZ9950100

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions