missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PELP1/MNAR Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Coactivator of estrogen receptor-mediated transcription and a corepressor of other nuclear hormone receptors and sequence- specific transcription factors. Plays a role in estrogen receptor (ER) genomic activity when present in the nuclear compartment by activating the ER target genes in a hormonal stimulation dependent manner. Can facilitate ER non-genomic signaling via SRC and PI3K interaction in the cytosol. Plays a role in E2-mediated cell cycle progression by interacting with RB1. May have important functional implications in ER/growth factor cross-talk. Interacts with several growth factor signaling components including EGFR and HRS. Involved in nuclear receptor signaling via its interaction with AR and NR3C1. May promote tumorigenesis via its interaction with and modulation of several oncogenes including SRC, PI3K, STAT3 and EGFR. Plays a role in cancer cell metastasis via its ability to modulate E2-mediated cytoskeleton changes and cell migration via its interaction with SRC and PI3K. Functions as the key stabilizing component of the Five Friends of Methylated CHTOP (5FMC) complex; the 5FMC complex is recruited to ZNF148 by methylated CHTOP, leading to desumoylation of ZNF148 and subsequent transactivation of ZNF148 target genes.
Spécifications
Spécifications
| Antigène | PELP1/MNAR |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Formule | TBS with 0.1% BSA and 0.09% sodium azide |
| Gène | PELP1 |
| Numéro d’acquisition du gène | Q8IZL8, Q9DBD5 |
| Alias du gène | HMX3, MNAR, P160, modulator of non-genomic activity of estrogen receptor; modulator of nongenomic activity of estrogen receptor; proline and glutamic acid rich nuclear protein; proline, glutamic acid and leucine rich protein 1; proline-, glutamic acid- and leucine-rich protein 1; proline-, glutamic acid-, leucine-rich protein 1; transcription factor HMX3 |
| Symboles de gène | Pelp1 |
| Espèces hôtes | Rabbit |
| Afficher plus de résultats |