missing translation for 'onlineSavingsMsg'
Learn More

Pelitinib 97.0+%, TCI America™

Catalog No. P252925MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P252925MG 25 mg
P2529100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P252925MG Supplier TCI America Supplier No. P252925MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 257933-82-7
Molecular Formula C24H23ClFN5O2
Molecular Weight (g/mol) 467.929
MDL Number MFCD09837868
InChI Key WVUNYSQLFKLYNI-AATRIKPKSA-N
Synonym EKB 569, (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-dimethylamino-2-butenamide
PubChem CID 6445562
ChEBI CHEBI:38927
IUPAC Name (E)-N-[4-(3-chloro-4-fluoroanilino)-3-cyano-7-ethoxyquinolin-6-yl]-4-(dimethylamino)but-2-enamide
SMILES CCOC1=C(C=C2C(=C1)N=CC(=C2NC3=CC(=C(C=C3)F)Cl)C#N)NC(=O)C=CCN(C)C

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 25 mg
UN Number 2811
Formula Weight 467.93
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pelitinib
TSCA No
Recommended Storage Freezer